7-Nitroindole
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:6960-42-5
- Formula:C₈H₆N₂O₂
- MDL Number:MFCD00005683
- Molecular Weight:162.15 g/mol
- Melting Pt:96…97 °C
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
- CAS Number:6960-42-5
- Formula:C₈H₆N₂O₂
- MDL Number:MFCD00005683
- Molecular Weight:162.15 g/mol
- Melting Pt:96…97 °C
- Storage Temperature:Ambient



