Order Entry
Export
Orders LinkContactUsLinkComponent
5-Bromophthalide

5-Bromophthalide

Bottle, 5 g
Catalog # APOSOR61443-5G
Irritant
Supplier:   Apollo Scientific
5-Bromophthalide
Catalog # APOSOR61443-5G
Supplier:  Apollo Scientific
CAS Number:  64169-34-2
5-Bromophthalide

Bottle, 5 g

CAS Number:  64169-34-2
Price
21,80 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  5-Bromoisobenzofuran-1(3H)-one
  • CAS Number:64169-34-2
  • Formula:C₈H₅BrO₂
  • MDL Number:MFCD01797360
  • Molecular Weight:213.03 g/mol
  • Melting Pt:162…166 °C
  • Storage Temperature:Ambient
  • EINECS:447-190-5
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications