Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,6-Dichloropyridin-3-amine
4,6-Dichloropyridin-3-amine 97+%
4,6-Dichloropyridin-3-amine 97+%
:
Apollo Scientific
4,6-Dichloropyridin-3-amine 97+%
:
Apollo Scientific
:
7321-93-9
:
:
C₅H₄Cl₂N₂
:
MFCD00234054
:
163.001 g/mol
:
80…85 °C
:
GHS07
:
Refrigerator