3-Mercapto-1,2-propanediol 97%
Catalog # APOSOR61438-10ML
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:10 mL
- Seq:2275
Specifications
About this item
:
:108.16 g/mol
Boiling Pt:118 °C/5mmHg
Density:1.247 g/cm³ (25 °C)
Flash Pt:113 °C
Storage Temperature:Refrigerator
EINECS:202-495-0
UN:2810
ADR:6.1,III
:GHS06
CAS Number:96-27-5
Formula:C₃H₈O₂S
MDL Number:MFCD00004879