Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Bis(2-methylstyryl)benzene
1,4-Bis(2-methylstyryl)benzene
1,4-Bis(2-methylstyryl)benzene
Irritant
1,4-Bis(2-methylstyryl)benzene
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1,4-Bis(2-methylstyryl)benzene
Supplier:
Apollo Scientific
About this item
CAS Number:
13280-61-0
:
Formula:
C₂₄H₂₂
MDL Number:
MFCD00008529
:
310.44 g/mol
Melting Pt:
180…182 °C
EINECS:
236-285-5
:
GHS07
Storage Temperature:
Ambient