Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Hydroxy-7-methoxycoumarin
4-Hydroxy-7-methoxycoumarin 98%
4-Hydroxy-7-methoxycoumarin 98%
Irritant
4-Hydroxy-7-methoxycoumarin 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Hydroxy-7-methoxycoumarin 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
17575-15-4
:
Formula:
C₁₀H₈O₄
MDL Number:
MFCD00673700
Melting Pt:
263…265 °C
:
192.17 g/mol
:
GHS07
Storage Temperature:
Ambient