4-Chlorophenylurea 98%
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 1-(4-Chlorophenyl)urea
- CAS Number:140-38-5
- Formula:C₇H₇ClN₂O
- MDL Number:MFCD00014788
- Molecular Weight:170.6 g/mol
- Melting Pt:205…205 °C
- Storage Temperature:Ambient
5 Options Available Below
Product Details & Documents
Synonyms: 1-(4-Chlorophenyl)urea
- CAS Number:140-38-5
- Formula:C₇H₇ClN₂O
- MDL Number:MFCD00014788
- Molecular Weight:170.6 g/mol
- Melting Pt:205…205 °C
- Storage Temperature:Ambient
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



