

3,5-Dibromo-2-hydroxypyrazine
Catalog # APOSOR61104-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:253.8800 g/mol
Melting Pt:174…178 °C
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:21943-15-7
Formula:C4H2Br2N2O
MDL Number:MFCD07371406