Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 6-chloropicolinate
Methyl-6-chloropicolinate
Methyl-6-chloropicolinate
Irritant
Methyl-6-chloropicolinate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Methyl-6-chloropicolinate
Supplier:
Apollo Scientific
About this item
CAS Number:
6636-55-1
:
Formula:
C₇H₆ClNO₂
MDL Number:
MFCD00155391
:
171.58 g/mol
Melting Pt:
93…94 °C
:
GHS07
Storage Temperature:
Refrigerator