Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Dimethylthiazole
2,5-Dimethylthiazole
2,5-Dimethylthiazole
2,5-Dimethylthiazole
APOSOR61055-5G
:
Apollo Scientific
:
2,5-Dimethylthiazole
APOSOR61055-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
113.1808 g/mol
:
148…153 °C
:
1.057 g/cm³ (25 °C)
:
GHS07
:
Refrigerator
:
4175-66-0
:
C₅H₇NS
:
MFCD00130120