Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dibromotoluene
2,6-Dibromotoluene
2,6-Dibromotoluene
2,6-Dibromotoluene
APOSOR60137-25G
:
Apollo Scientific
:
2,6-Dibromotoluene
APOSOR60137-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
249.93 g/mol
:
112…113 °C/7mmHg
:
44349 °C
:
1.812 g/cm³ (25 °C)
:
GHS07
:
Ambient
:
69321-60-4
:
C₇H₆Br₂
:
MFCD00013524