
6-Chloro-3,4-pyridinediamine
Catalog # APOSOR60103-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:143.5700 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:89182-17-2
Formula:C₅H₆ClN₃
MDL Number:MFCD04037269