Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dichloro-p-benzoquinone
2,6-Dichloro-p-benzoquinone 95%
2,6-Dichloro-p-benzoquinone 95%
2,6-Dichloro-p-benzoquinone 95%
Supplier:
Apollo Scientific
Certificates
Expand image
2,6-Dichloro-p-benzoquinone 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
697-91-6
:
Formula:
C₆H₂Cl₂O₂
MDL Number:
MFCD00037159
:
176.99 g/mol
Melting Pt:
122…124 °C
Storage Temperature:
Ambient