Order Entry
Export
ContactUsLinkComponent
 
Irritant
2,3-Dibromoisonicotinic acid 97%
Catalog # APOSOR59813-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,3-Dibromoisonicotinic acid 97%
Catalog # APOSOR59813-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :280.90 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1020056-98-7
Formula:C₆H₃Br₂NO₂
MDL Number:MFCD09997708