
2,6-Dichlorobenzylamine
Catalog # APOSOR59807-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:176.04 g/mol
Storage Temperature:Ambient
UN:2735
ADR:8,III
:GHS05
CAS Number:6575-27-5
Formula:C₇H₇Cl₂N
MDL Number:MFCD00047928