Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 3,3-dimethoxypropionate
Methyl-3,3-dimethoxypropionate 98%
Methyl-3,3-dimethoxypropionate 98%
:
Apollo Scientific
Methyl-3,3-dimethoxypropionate 98%
:
Apollo Scientific
:
7424-91-1
:
:
C₆H₁₂O₄
:
MFCD00010650
:
148.16 g/mol
:
76…77 °C/20mmHg
:
1.045 g/cm³ (25 °C)
:
66 °C
:
231-055-0
:
GHS07
:
Ambient