Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Imidazo[1,2-a]pyridine-7-carboxylic acid
Imidazo[1,2-a]pyridine-7-carboxylic acid
Imidazo[1,2-a]pyridine-7-carboxylic acid
Irritant
Imidazo[1,2-a]pyridine-7-carboxylic acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Imidazo[1,2-a]pyridine-7-carboxylic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
648423-85-2
:
Formula:
C₈H₆N₂O₂
MDL Number:
MFCD09834781
:
162.14 g/mol
Melting Pt:
244…247 °C
:
GHS07
Storage Temperature:
Ambient