Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,4-Dimethyl-2-cyclohexen-1-one
4,4-Dimethyl-2-cyclohexen-1-one 98%
4,4-Dimethyl-2-cyclohexen-1-one 98%
:
Apollo Scientific
4,4-Dimethyl-2-cyclohexen-1-one 98%
:
Apollo Scientific
:
1073-13-8
:
:
C₈H₁₂O
:
MFCD00009695
:
124.18 g/mol
:
72…74 °C/20mmHg
:
0.944 g/cm³ (25 °C)
:
64 °C
:
GHS07
:
Ambient