
2-Bromo-m-xylene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:576-22-7
:
Formula:C₈H₉Br
MDL Number:MFCD00000075
:185.06 g/mol
Boiling Pt:205…206 °C
Melting Pt:–10 °C
Density:1.389 g/cm³ (25 °C)
Flash Pt:73 °C
EINECS:209-397-7
:GHS07
Storage Temperature:Ambient