Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-2,6-dichloropyridine
3-Bromo-2,6-dichloropyridine 97%
3-Bromo-2,6-dichloropyridine 97%
:
Apollo Scientific
3-Bromo-2,6-dichloropyridine 97%
:
Apollo Scientific
:
866755-20-6
:
:
C₅H₂BrCl₂N
:
MFCD06798231
:
226.89 g/mol
:
68…71 °C
:
GHS07
:
Ambient