Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Phenyl(4,6-dimethoxypyrimidin-2-yl)carbamate
Phenyl(4,6-dimethoxypyrimidin-2-yl)carbamate
Phenyl(4,6-dimethoxypyrimidin-2-yl)carbamate
Irritant
Phenyl(4,6-dimethoxypyrimidin-2-yl)carbamate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Phenyl(4,6-dimethoxypyrimidin-2-yl)carbamate
Supplier:
Apollo Scientific
About this item
CAS Number:
89392-03-0
:
Formula:
C13H13N3O4
MDL Number:
MFCD02947731
:
275.2600 g/mol
:
GHS07
Storage Temperature:
Ambient