Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Dibromo-o-xylene
4,5-Dibromo-o-xylene 98+%
4,5-Dibromo-o-xylene 98+%
Irritant
4,5-Dibromo-o-xylene 98+%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4,5-Dibromo-o-xylene 98+%
Supplier:
Apollo Scientific
About this item
CAS Number:
24932-48-7
:
Formula:
C₈H₈Br₂
MDL Number:
MFCD00082743
:
263.96 g/mol
Boiling Pt:
278 °C
Melting Pt:
85…87 °C
:
GHS07
Storage Temperature:
Ambient