
3-Bromo-5-nitrotoluene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:52488-28-5
:
Formula:C₇H₆BrNO₂
MDL Number:MFCD06659815
:216.03 g/mol
Boiling Pt:269…270 °C
Melting Pt:81…84 °C
Storage Temperature:Ambient
EINECS:257-955-3
:GHS07