Order Entry
Export
ContactUsLinkComponent
2,5-Dibromo-1-tritylimidazole 95%
2,5-Dibromo-1-tritylimidazole 95%
Catalog # APOSOR5621-1G
Supplier:  Apollo Scientific
CAS Number:  
2,5-Dibromo-1-tritylimidazole 95%
Catalog # APOSOR5621-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:154…158 °C
Storage Temperature:Refrigerator
CAS Number:850429-53-7
Formula:C₂₂H₁₆Br₂N₂
MDL Number:MFCD04115731