
Imidazole-4-carboxylic acid ethyl ester 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:23785-21-9
:MFCD02646639
Formula:C₆H₈N₂O₂
MDL Number:MFCD07363812
:140.14 g/mol
Melting Pt:160…162 °C
:GHS07
Storage Temperature:Refrigerator