Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Hydroxy-2-nitropyridine
3-Hydroxy-2-nitropyridine 98%
3-Hydroxy-2-nitropyridine 98%
Irritant
3-Hydroxy-2-nitropyridine 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3-Hydroxy-2-nitropyridine 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
15128-82-2
:
Formula:
C₅H₄N₂O₃
MDL Number:
MFCD00006258
:
140.1 g/mol
Melting Pt:
69…73 °C
EINECS:
239-191-2
:
GHS07
Storage Temperature:
Ambient