Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Dibromo-2-iodobenzene
1,4-Dibromo-2-iodobenzene 99%
1,4-Dibromo-2-iodobenzene 99%
Irritant
1,4-Dibromo-2-iodobenzene 99%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1,4-Dibromo-2-iodobenzene 99%
Supplier:
Apollo Scientific
About this item
CAS Number:
89284-52-6
:
Formula:
C₆H₃Br₂I
MDL Number:
MFCD07778993
:
361.80 g/mol
Melting Pt:
38…39 °C
:
GHS07
Storage Temperature:
Ambient