Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Dihydroxynaphthalene
1,3-Dihydroxynaphthalene 99%
1,3-Dihydroxynaphthalene 99%
:
Apollo Scientific
1,3-Dihydroxynaphthalene 99%
:
Apollo Scientific
:
132-86-5
:
:
C₁₀H₈O₂
:
MFCD00003965
:
160.17 g/mol
:
123…126 °C
:
GHS07
:
Ambient