Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,3-Benzenedithiol

Corrosive
Irritant
Supplier:   Apollo Scientific
1,3-Benzenedithiol
Supplier:  Apollo Scientific
Certificates
1,3-Benzenedithiol

4 Options Available Below

Product Details & Documents
Synonyms:  Benzene-1,3-dithiol
  • CAS Number:626-04-0
  • Formula:C₆H₆S₂
  • MDL Number:MFCD00004842
  • Molecular Weight:142.25 g/mol
  • Boiling Pt:244 °C
  • Melting Pt:24…25 °C
  • Density:1.24 g/cm³ (25 °C)
  • Flash Pt:113 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 43,50 €
    43,50 €
    1 * 250 mg
       43,50 €

    Specifications:
    New Product
    New Product
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 94,10 €
    94,10 €
    1 * 1 g
       94,10 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 362,00 €
    362,00 €
    1 * 5 g
       362,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 1 730,00 €
    1 730,00 €
    1 * 25 g
       1 730,00 €

    Specifications:
    New Product
    New Product