Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
8-Bromo[1,2,4]triazolo[1,5-a]pyridine
8-Bromo[1,2,4]triazolo[1,5-a]pyridine 95%
8-Bromo[1,2,4]triazolo[1,5-a]pyridine 95%
:
Apollo Scientific
8-Bromo[1,2,4]triazolo[1,5-a]pyridine 95%
:
Apollo Scientific
:
868362-18-9
:
:
C₆H₄BrN₃
:
MFCD10699199
:
198.03 g/mol
:
152…157 °C
:
GHS07
:
Ambient