Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-(1-Naphthyl)formamide
N-(1-Naphthyl)formamide 99%
N-(1-Naphthyl)formamide 99%
Irritant
N-(1-Naphthyl)formamide 99%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
N-(1-Naphthyl)formamide 99%
Supplier:
Apollo Scientific
About this item
CAS Number:
6330-51-4
:
Formula:
C₁₁H₉NO
MDL Number:
MFCD00086455
:
171.20 g/mol
Boiling Pt:
140 °C
:
GHS07
Storage Temperature:
Ambient