Order Entry
Export
ContactUsLinkComponent
4-Bromo-2,6-xylenol 98%
Irritant
4-Bromo-2,6-xylenol 98%
Catalog # APOSOR5479-10G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Bromo-2,6-xylenol 98%
Catalog # APOSOR5479-10G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :201.06 g/mol
Melting Pt:74…79 °C
EINECS:219-153-1
 :GHS07
Storage Temperature:Ambient
CAS Number:2374-05-2
Formula:C₈H₉BrO
MDL Number:MFCD00002314