
N-Methyl-3-pyrrolidinol
Catalog # APOSOR5467-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:101.15 g/mol
Boiling Pt:50…52 °C/1mmHg
Density:0.921 g/cm³ (25 °C)
Flash Pt:71 °C
EINECS:236-189-3
:GHS07
Storage Temperature:Ambient
CAS Number:13220-33-2
Formula:C₅H₁₁NO
MDL Number:MFCD00003176