Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,3-Diaminophenazine, Technical Grade

Irritant
Supplier:   Apollo Scientific
2,3-Diaminophenazine, Technical Grade
Supplier:  Apollo Scientific
Certificates
2,3-Diaminophenazine, Technical Grade

2 Options Available Below

Product Details & Documents
Synonyms:  DAP, Phenazine-2,3-diyldiamine
  • CAS Number:655-86-7
  • Formula:C₁₂H₁₀N₄
  • MDL Number:MFCD00160693
  • Molecular Weight:210.24 g/mol
  • Melting Pt:>300 °C
  • Melting Pt:>300 °C
  • Storage Temperature:Ambient
  • EINECS:211-512-0
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 43,50 €
    43,50 €
    1 * 1 g
       43,50 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 167,00 €
    167,00 €
    1 * 5 g
       167,00 €