Order Entry
Export
ContactUsLinkComponent
4,6-Dihydrothieno[3,4-b]thiophene-2-carboxylic acid 96%
Irritant
4,6-Dihydrothieno[3,4-b]thiophene-2-carboxylic acid 96%
Catalog # APOSOR54533-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4,6-Dihydrothieno[3,4-b]thiophene-2-carboxylic acid 96%
Catalog # APOSOR54533-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
Melting Pt:240 °C
 :186.24 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:7712-05-2
Formula:C₇H₆O₂S₂
MDL Number:MFCD20642620