Order Entry
Export
Orders LinkContactUsLinkComponent
1-Naphthol 99%

1-Naphthol 99%

Toxic
Corrosive
Irritant
Supplier:   Apollo Scientific
1-Naphthol 99%
Supplier:  Apollo Scientific
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  1-Hydroxynaphthalene, α-Naphthol, α-Hydroxynaphthalene
  • CAS Number:90-15-3
  • Formula:C₁₀H₈O
  • MDL Number:MFCD00003930
  • Molecular Weight:144.17 g/mol
  • Boiling Pt:278…280 °C
  • Melting Pt:95…97 °C
  • Density:1.28 g/cm³ (25 °C)
  • Flash Pt:125 °C
  • Storage Temperature:Ambient
  • UN:2811
  • ADR:6.1,III

Documents

Product Family Options

Product Information