(1-Ethoxycyclopropoxy)trimethylsilane
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:27374-25-0
:
Formula:C₈H₁₈O₂Si
MDL Number:MFCD00074986
:174.315 g/mol
Boiling Pt:50…53 °C
Density:0.867 g/cm³ (25 °C)
Flash Pt:26 °C
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS02
:GHS07