
2,4'-Dichlorobenzophenone 98%
Catalog # APOSOR54515-500G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:251.11 g/mol
Melting Pt:63…66 °C
:GHS07
Storage Temperature:Ambient
CAS Number:85-29-0
Formula:C₁₃H₈Cl₂O
MDL Number:MFCD00038744