Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 2-methoxynicotinate
Methyl-2-methoxynicotinate
Methyl-2-methoxynicotinate
Irritant
Methyl-2-methoxynicotinate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Methyl-2-methoxynicotinate
Supplier:
Apollo Scientific
About this item
CAS Number:
67367-26-4
:
Formula:
C₈H₉NO₃
MDL Number:
MFCD06801052
:
167.16 g/mol
Melting Pt:
25…27 °C
:
GHS07
Storage Temperature:
Refrigerator