Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,1'-Thiocarbonyldiimidazole
1,1'-Thiocarbonyldiimidazole 95%
1,1'-Thiocarbonyldiimidazole 95%
:
Apollo Scientific
1,1'-Thiocarbonyldiimidazole 95%
:
Apollo Scientific
:
6160-65-2
:
:
C₇H₆N₄S
:
MFCD00005289
:
178.22 g/mol
:
88…105 °C
:
GHS07
:
Refrigerator