
5,6-Dimethoxyindole 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:14430-23-0
:
Formula:C₁₀H₁₁NO₂
MDL Number:MFCD00005675
:177.2 g/mol
Boiling Pt:198 °C/8mmHg
Melting Pt:154…157 °C
:GHS07
Storage Temperature:Refrigerator