1,1-Cyclobutanedicarboxylic acid 98%
Catalog # APOSOR5361-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:144.13 g/mol
Melting Pt:158 °C
Storage Temperature:Ambient
CAS Number:5445-51-2
Formula:C₆H₈O₄
MDL Number:MFCD00001325