Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Lutidine
2,6-Lutidine 98+%
2,6-Lutidine 98+%
:
Apollo Scientific
2,6-Lutidine 98+%
:
Apollo Scientific
:
108-48-5
:
:
C₇H₉N
:
MFCD00006345
:
107.16 g/mol
:
143…145 °C
:
–6 °C
:
0.921 g/cm³ (25 °C)
:
33 °C
:
Refrigerator
:
203-587-3
:
1993
:
3,III
:
GHS02
:
GHS07