Order Entry
Export
Orders LinkContactUsLinkComponent
2,5-Dimethylindole

2,5-Dimethylindole

Corrosive
Irritant
Supplier:   Apollo Scientific
2,5-Dimethylindole
Supplier:  Apollo Scientific
Certificates
2,5-Dimethylindole

3 Options Available Below

Product Details & Documents
Synonyms:  2,5-Dimethyl-1H-indole
  • CAS Number:1196-79-8
  • Formula:C₁₀H₁₁N
  • MDL Number:MFCD00005621
  • Molecular Weight:145.2 g/mol
  • Melting Pt:113…114 °C
  • Storage Temperature:Ambient

Documents

Product Family Options

Product Information