Order Entry
Export
ContactUsLinkComponent
9,10-Dihydro-9-oxa-10-phosphaphenanthrene-10-oxide 95+%
9,10-Dihydro-9-oxa-10-phosphaphenanthrene-10-oxide 95+%
Catalog # APOSOR53111-100G
Supplier:  Apollo Scientific
CAS Number:  
9,10-Dihydro-9-oxa-10-phosphaphenanthrene-10-oxide 95+%
Catalog # APOSOR53111-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :216.18 g/mol
Melting Pt:119 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:35948-25-5
Formula:C₁₂H₉O₂P
MDL Number:MFCD00040561