Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,7-Heptanediol
1,7-Heptanediol 98%
1,7-Heptanediol 98%
:
Apollo Scientific
1,7-Heptanediol 98%
:
Apollo Scientific
:
629-30-1
:
:
C₇H₁₆O₂
:
MFCD00002987
:
132.2 g/mol
:
259 °C
:
17…19 °C
:
0.95 g/cm³ (25 °C)
:
113 °C
:
GHS07
:
Ambient