Order Entry
Export
ContactUsLinkComponent
Pyrazine-2,5-dicarboxylic acid 97%
Irritant
Pyrazine-2,5-dicarboxylic acid 97%
Catalog # APOSOR52999-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Pyrazine-2,5-dicarboxylic acid 97%
Catalog # APOSOR52999-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :168.108 g/mol
Melting Pt:255…260 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:122-05-4
Formula:C₆H₄N₂O₄
MDL Number:MFCD01630902