1-Chloro-N,N-2-trimethylpropenylamine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:26189-59-3
:
Formula:C₆H₁₂ClN
MDL Number:MFCD00800562
:133.619 g/mol
Boiling Pt:129…130 °C
Density:1.01 g/cm³ (25 °C)
Flash Pt:26.67 °C
Storage Temperature:Freezer
UN:2734
ADR:8+3,II