
1,5,7-Triazabicyclo[4.4.0]dec-5-ene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5807-14-7
:
Formula:C₇H₁₃N₃
MDL Number:MFCD00043003
Melting Pt:125…130 °C
UN:3263
ADR:8,III
:139.20 g/mol
:GHS05
Storage Temperature:Ambient