Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Bromopyrazolo[1,5-a]pyridine
5-Bromopyrazolo[1,5-a]pyridine 97%
5-Bromopyrazolo[1,5-a]pyridine 97%
Irritant
5-Bromopyrazolo[1,5-a]pyridine 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5-Bromopyrazolo[1,5-a]pyridine 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
1060812-84-1
:
Formula:
C₇H₅BrN₂
MDL Number:
MFCD13189494
:
197.032 g/mol
:
GHS07
Storage Temperature:
Refrigerator